Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018819.4 | NP_061289.1 | 4, 5 | 375 | ||
GenBank mRNA | AF181116.1 | - | 4, 5 | 331 | ||
AK002241.1 | - | 4, 5 | 376 | |||
AK002266.1 | - | 4, 5 | 354 | |||
AK004013.1 | - | 4, 5 | 331 | |||
AK005348.1 | - | 4, 5 | 367 | |||
AK013391.1 | - | 4, 5 | 376 | |||
AK014421.1 | - | 4, 5 | 333 | |||
AK014435.1 | - | 4, 5 | 605 | |||
AK135943.1 | - | 4, 5 | 346 | |||
AK170514.1 | - | 4, 5 | 392 | |||
BC024365.1 | - | 4, 5 | 337 | 4988147 | ||
BC103767.1 | - | 4, 5 | 378 | 30840199 |