Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc23a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018824.2 | NP_061294.2 | 2 | 336 | ||
GenBank mRNA | AB038145.1 | - | 1 | 72 | ||
AK087175.1 | - | 2 | 344 | |||
AK169991.1 | - | 2 | 338 | |||
AY004874.1 | - | 2 | 290 | |||
BC050823.1 | - | 1 | 100 | 6332451 |