Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018830.1 | NP_061300.1 | 8 | 974 | ||
GenBank mRNA | AB037111.1 | - | 9 | 1259 | ||
AB037181.1 | - | 8 | 974 | |||
AK034493.1 | - | 8 | 1205 | |||
AK041937.1 | - | 8 | 1163 | |||
AK046540.1 | - | 8 | 1168 | |||
AK047692.1 | - | 8 | 1162 | |||
AK080951.1 | - | 8 | 1151 | |||
AK166100.1 | - | 8 | 1222 | |||
BC137898.1 | - | 8 | 957 | 8860917 | ||
BC145016.1 | - | 8 | 960 | 9053564 |