Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018832.2 | NP_061302.2 | 6 | 1163 | ||
GenBank mRNA | AF229645.1 | - | 6 | 1183 | ||
AK077898.1 | - | 6 | 1130 | |||
AK158091.1 | - | 6 | 1103 | |||
BC099459.1 | - | 6 | 1163 | 30847792 | ||
BC132580.1 | - | 6 | 1102 | 40130857 | ||
BC132582.1 | - | 6 | 1102 | 40130859 |