Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018864.5 | NP_061352.2 | 6, 7 | 582 | ||
GenBank mRNA | AF042730.1 | - | 6, 7 | 527 | ||
AK011770.1 | - | 6, 7 | 520 | |||
AK078406.1 | - | 6, 7 | 531 | |||
AK083238.1 | - | 6, 7 | 516 | |||
AK145472.1 | - | 519 | ||||
AK147778.1 | - | 6, 7 | 542 | |||
AK152774.1 | - | 6, 7 | 533 | |||
AK153457.1 | - | 6, 7 | 549 | |||
AK160203.1 | - | 6, 7 | 520 | |||
AK160467.1 | - | 6, 7 | 514 | |||
AK165977.1 | - | 6, 7 | 524 | |||
AK168457.1 | - | 6, 7 | 557 | |||
AK179584.1 | - | 1, 2 | 66 | |||
AK189530.1 | - | 1, 2 | 66 | |||
AK210121.1 | - | 1, 2 | 67 | |||
BC049080.1 | - | 6, 7 | 523 | 6419835 | ||
BC055722.1 | - | 5, 6 | 477 | 6413389 |