Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018864.5 | NP_061352.2 | 9 | 2134 | ||
GenBank mRNA | AF042730.1 | - | 9 | 2080 | ||
AK011770.1 | - | 9 | 2072 | |||
AK078406.1 | - | 9 | 2082 | |||
AK083238.1 | - | 9 | 2069 | |||
AK145472.1 | - | 2073 | ||||
AK147778.1 | - | 9 | 2094 | |||
AK160203.1 | - | 9 | 2072 | |||
AK160467.1 | - | 9 | 2067 | |||
BC049080.1 | - | 8 | 2876 | 6419835 | ||
BC055722.1 | - | 8 | 2029 | 6413389 |