Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018872.2 | NP_061360.2 | 9, 10 | 1123 | ||
GenBank mRNA | AB093221.1 | - | 5, 6 | 559 | ||
AF060565.1 | - | 9, 10 | 825 | |||
AK139317.1 | - | 9, 10 | 1125 |