Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285870.1 | NP_001272799.1 | 5 | 900 | ||
NM_001285871.1 | NP_001272800.1 | 5 | 1085 | |||
NM_001285873.1 | NP_001272802.1 | 5 | 900 | |||
NM_018880.3 | NP_061368.1 | 5 | 874 | |||
GenBank mRNA | AB030912.1 | - | 5 | 901 | ||
AF220019.1 | - | 5 | 793 | |||
AK019165.1 | - | 5 | 835 | |||
AK144674.1 | - | 5 | 803 | |||
AK170029.1 | - | 5 | 838 | |||
BC003347.1 | - | 5 | 915 | |||
BC034263.1 | - | 5 | 839 | 4503884 |