Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285870.1 | NP_001272799.1 | 9 | 1990 | ||
NM_001285871.1 | NP_001272800.1 | 9 | 2175 | |||
NM_001285873.1 | NP_001272802.1 | 9 | 1990 | |||
NM_018880.3 | NP_061368.1 | 9 | 1964 | |||
GenBank mRNA | AB030912.1 | - | 9 | 1991 | ||
AF220019.1 | - | 9 | 1883 | |||
AK019165.1 | - | 9 | 1925 | |||
AK144674.1 | - | 9 | 1893 | |||
AK170029.1 | - | 9 | 1928 | |||
AK189185.1 | - | 1 | 41 | |||
BC034263.1 | - | 9 | 1929 | 4503884 |