Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018881.3 | NP_061369.2 | 5 | 549 | ||
GenBank mRNA | AK129026.1 | - | 1 | 82 | ||
BC031415.1 | - | 5 | 549 | 3990305 |