Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019397.3 | NP_062270.1 | 3, 4 | 464 | ||
GenBank mRNA | AJ245672.1 | - | 3, 4 | 466 | ||
AK053738.1 | - | 3, 4 | 461 | |||
AK078534.1 | - | 3, 4 | 442 | |||
BC117702.2 | - | 3, 4 | 450 | 40062530 |