Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019403.3 | NP_062276.1 | 1 | 167 | ||
GenBank mRNA | AK012729.1 | - | 1 | 159 | ||
AK019289.1 | - | 1 | 178 | |||
AK020514.2 | - | 1 | 168 | |||
AK020515.2 | - | 1 | 186 | |||
BC012031.1 | - | 1 | 118 | |||
BC016449.1 | - | 1 | 167 | 4191746 |