Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORF4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001265604.1 | NP_001252533.1 | 3 | 294 | ||
NM_001265605.1 | NP_001252534.1 | 3 | 194 | |||
NM_006791.3 | NP_006782.1 | 3 | 341 | |||
NM_206839.2 | NP_996670.1 | 3 | 341 | |||
XM_017021872.1 | XP_016877361.1 | 3 | 306 | |||
GenBank mRNA | AF070664.1 | - | 3 | 148 | ||
AF100615.1 | - | 3 | 235 | |||
AF109188.1 | - | 3 | 130 | |||
AF161546.1 | - | 3 | 127 | |||
AF167173.1 | - | 3 | 143 | |||
AF218011.1 | - | 3 | 246 | |||
AK095823.1 | - | 4 | 743 | |||
AK098485.1 | - | 3 | 261 | |||
AK296650.1 | - | 3 | 232 | |||
AK300789.1 | - | 3 | 278 | |||
AK310181.1 | - | 2 | 79 | |||
AK310592.1 | - | 3 | 140 | |||
AK310702.1 | - | 3 | 149 | |||
AL137697.1 | - | 3 | 135 | |||
AY148481.1 | - | 3 | 235 | |||
BC002936.2 | - | 3 | 222 | 3955120 | ||
BC022845.2 | - | 3 | 222 | 2988803 | ||
BC067826.1 | - | 3 | 117 | 4824022 | ||
BC141840.1 | - | 3 | 149 | 8860303 | ||
BG771854.1 | - | 3 | 351 | |||
BX382045.2 | - | 3 | 138 | |||
DB455429.1 | - | 3 | 194 |