Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hgfac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019447.2 | NP_062320.2 | 9 | 1108 | ||
GenBank mRNA | AF099017.1 | - | 9 | 1068 | ||
AF224724.1 | - | 9 | 1068 | |||
AK007987.1 | - | 9 | 1106 | |||
AK075718.1 | - | 9 | 1017 | |||
AK144554.1 | - | 9 | 1079 | |||
AK149522.1 | - | 9 | 1072 | |||
AK165433.1 | - | 7 | 1418 | |||
AK169989.1 | - | 9 | 1104 | |||
BC019376.1 | - | 9 | 1073 | 5134851 |