Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnmt3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081695.2 | NP_001075164.1 | 3, 4 | 396 | ||
NM_001284197.1 | NP_001271126.1 | 1, 2 | 237 | |||
NM_019448.4 | NP_062321.1 | 2, 3 | 250 | |||
GenBank mRNA | AF220524.1 | - | 1, 2 | 181 | ||
AJ404467.1 | - | 1, 2 | 222 | |||
AK010434.1 | - | 2, 3 | 251 | |||
AV473569.1 | - | 1, 2 | 237 | |||
BC083147.1 | - | 1, 2 | 235 | 30447906 | ||
EF051624.1 | - | 3, 4 | 396 |