Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081549.2 | NP_001075018.1 | 4 | 1796 | ||
NM_019466.4 | NP_062339.2 | 4 | 1642 | |||
GenBank mRNA | AF260717.1 | - | 4 | 1566 | ||
AF263239.1 | - | 4 | 1582 | |||
AF263240.1 | - | 4 | 1665 | |||
AK003336.1 | - | 1 | 514 | |||
AK146764.1 | - | 4 | 1771 | |||
AY325904.1 | - | 4 | 1752 | |||
BC013551.1 | - | 4 | 1621 | 4236038 |