Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006808.2 | NP_006799.1 | 2, 3 | 185 | ||
GenBank mRNA | BC001734.1 | - | 2, 3 | 185 | 3502133 | |
CR456883.1 | - | 1, 2 | 109 | |||
CR542159.1 | - | 1, 2 | 109 | |||
L25085.1 | - | 2, 3 | 172 |