Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019538.4 | NP_062411.1 | 3 | 741 | ||
GenBank mRNA | AF234653.1 | - | 3 | 742 | ||
AF250838.1 | - | 3 | 738 | |||
AK005501.1 | - | 3 | 742 | |||
AK145876.1 | - | 3 | 707 | |||
BC051666.1 | - | 3 | 727 | 30017089 |