Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prodh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019546.5 | NP_062419.2 | 10 | 1237 | ||
GenBank mRNA | AF222851.1 | - | 10 | 1224 | ||
AK011020.1 | - | 10 | 1229 | |||
AK050141.1 | - | 10 | 1238 | |||
BC018182.1 | - | 10 | 1223 | 4161894 | ||
BC052815.1 | - | 4 | 375 | |||
U80019.1 | - | 10 | 1235 |