Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019550.2 | NP_062423.1 | 14 | 2051 | ||
GenBank mRNA | AF095718.2 | - | 14 | 1923 | ||
AK013734.1 | - | 3 | 611 | |||
AK033964.1 | - | 7 | 2455 | |||
AK135738.1 | - | 14 | 2051 | |||
AK137348.1 | - | 14 | 1929 | |||
BC010255.1 | - | 14 | 1933 | 3709255 |