Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019552.2 | NP_062425.1 | 3 | 1017 | ||
GenBank mRNA | AF266284.1 | - | 3 | 747 | ||
AK081782.1 | - | 3 | 729 | |||
BC046818.1 | - | 3 | 973 | 5702696 | ||
BC053020.1 | - | 3 | 973 | 5702696 | ||
BC054793.1 | - | 3 | 1017 | 6833785 |