Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rev1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019570.3 | NP_062516.2 | 18 | 3211 | ||
GenBank mRNA | AB057418.1 | - | 17 | 3000 | ||
AF179302.1 | - | 17 | 3000 | |||
BC006721.1 | - | 13 | 2535 | |||
BC058093.1 | - | 18 | 3211 | 6827108 |