Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019571.5 | NP_062517.1 | 8 | 1498 | ||
GenBank mRNA | AF121344.1 | - | 8 | 1513 | ||
AK013350.1 | - | 8 | 1464 | |||
AK033543.1 | - | 8 | 1197 | |||
AK182422.1 | - | 1 | 55 | |||
AK190582.1 | - | 1 | 90 | |||
AK195572.1 | - | 1 | 89 | |||
BC046784.1 | - | 8 | 1337 | 6416107 | ||
BC058695.1 | - | 8 | 1469 | 6813872 |