Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Patz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253690.1 | NP_001240619.1 | 5 | 2156 | ||
NM_001253691.1 | NP_001240620.1 | 5 | 2175 | |||
NM_019574.3 | NP_062520.1 | 4 | 2083 | |||
GenBank mRNA | AB029397.1 | - | 4 | 2083 | ||
AK018372.1 | - | 4 | 837 | |||
AK206872.1 | - | 1 | 30 | |||
AK217477.1 | - | 1 | 30 | |||
BC026673.1 | - | 4 | 754 | |||
BC043035.1 | - | 3 | 1909 | 5697155 | ||
BC055947.1 | - | 6 | 887 | |||
CA327687.1 | - | 5 | 763 |