Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Becn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019584.3 | NP_062530.2 | 8 | 875 | ||
GenBank mRNA | AF077302.2 | - | 7 | 737 | ||
AK028851.1 | - | 6 | 2317 | |||
AK031501.1 | - | 7 | 1502 | |||
AK032176.1 | - | 876 | ||||
AK050329.1 | - | 8 | 876 | |||
AK077392.1 | - | 8 | 962 | |||
AK083800.1 | - | 8 | 865 | |||
AK151531.1 | - | 8 | 842 | |||
AK214094.1 | - | 1 | 53 | |||
AK216278.1 | - | 3 | 340 | |||
AK217279.1 | - | 1 | 53 | |||
BC005770.1 | - | 8 | 856 | 2650059 |