Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006826.3 | NP_006817.1 | 2 | 384 | ||
GenBank mRNA | AK297624.1 | - | 3 | 217 | ||
AK302208.1 | - | 2 | 90 | |||
AK309462.1 | - | 3 | 213 | |||
AK316158.1 | - | 3 | 212 | |||
BC001197.1 | - | 321 | ||||
BC050601.1 | - | 1 | 197 | 6501365 | ||
BC056867.1 | - | 2 | 319 | 6164592 | ||
BC093019.1 | - | 2 | 326 | 30520472 | ||
BT020014.1 | - | 1 | 187 | |||
HY044745.1 | - | 2 | 392 | |||
X56468.1 | - | 2 | 312 | |||
X57347.1 | - | 2 | 287 |