Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019648.3 | NP_062622.1 | 12 | 2036 | ||
GenBank mRNA | AB003144.1 | - | 11 | 1347 | ||
AF434712.1 | - | 11 | 1426 | |||
AK053691.1 | - | 3 | 1417 | |||
AK076804.1 | - | 11 | 1379 | |||
AK090323.1 | - | 11 | 1365 | |||
AK147097.1 | - | 11 | 1471 | |||
AK159650.1 | - | 11 | 1450 | |||
AK163318.1 | - | 11 | 1453 | |||
AK165301.1 | - | 12 | 2038 | |||
AK165611.1 | - | 2 | 214 | |||
AK165845.1 | - | 11 | 1362 | |||
AK194694.1 | - | 1 | 172 | |||
BC002213.1 | - | 11 | 1448 | 3490341 | ||
BC010263.1 | - | 11 | 1420 | |||
BC092052.1 | - | 11 | 1413 | 3670685 |