Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnj1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168354.1 | NP_001161826.1 | 2 | 403 | ||
NM_019659.3 | NP_062633.1 | 2 | 401 | |||
GenBank mRNA | AF012834.1 | - | 2 | 222 | ||
AK085262.1 | - | 2 | 327 | |||
BC020525.1 | - | 2 | 403 | 4241262 | ||
CF168846.1 | - | 2 | 294 |