Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ykt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019661.4 | NP_062635.2 | 6, 7 | 698 | ||
GenBank mRNA | AF076956.1 | - | 6, 7 | 570 | ||
AK002916.1 | - | 6, 7 | 676 | |||
AK007486.1 | - | 6, 7 | 666 | |||
AK147098.1 | - | 6, 7 | 658 | |||
AK150472.1 | - | 6, 7 | 655 | |||
AK159253.1 | - | 6, 7 | 645 | |||
AK211548.1 | - | 212 | ||||
BC006760.1 | - | 6, 7 | 624 | 3257673 |