Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019667.2 | NP_062641.1 | 9 | 921 | ||
GenBank mRNA | AB012611.1 | - | 9 | 936 | ||
AK004604.2 | - | 9 | 921 | |||
AK035789.1 | - | 974 | ||||
AK044230.1 | - | 8 | 850 | |||
AK159699.1 | - | 9 | 950 | |||
AK168729.1 | - | 9 | 953 | |||
BC013818.1 | - | 9 | 957 | 3671426 |