Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019670.1 | NP_062644.1 | 5 | 512 | ||
GenBank mRNA | AF094519.1 | - | 5 | 512 | ||
AK137073.1 | - | 4 | 601 | |||
AK138694.1 | - | 5 | 542 | |||
AK161737.1 | - | 4 | 530 | |||
AK166882.1 | - | 5 | 568 | |||
AK220493.1 | - | 5 | 576 | |||
BC003250.1 | - | 4 | 546 | |||
BC085191.1 | - | 5 | 622 | 30693873 | ||
BC086779.1 | - | 5 | 587 |