Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047159.2 | NP_001040624.1 | 2 | 305 | ||
NM_019671.3 | NP_062645.2 | 4 | 426 | |||
GenBank mRNA | AF094520.1 | - | 4 | 322 | ||
AJ010045.1 | - | 2 | 268 | |||
AK082128.1 | - | 2 | 302 | |||
AK088632.1 | - | 2 | 269 | |||
AK139921.1 | - | 2 | 269 | |||
BC004699.1 | - | 2 | 279 | 3499258 | ||
CF167482.1 | - | 4 | 410 | |||
CN702942.1 | - | 2 | 166 |