Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stmn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019675.2 | NP_062649.1 | 3, 4 | 233 | ||
GenBank mRNA | AF105222.1 | - | 2, 3 | 205 | ||
BC046752.1 | - | 3, 4 | 233 | 5693069 |