Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019680.2 | NP_062654.2 | 9 | 2895 | ||
GenBank mRNA | AF016714.1 | - | 8 | 2831 |