Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019685.2 | NP_062659.1 | 6 | 723 | ||
GenBank mRNA | AF100694.1 | - | 6 | 762 | ||
AK012170.1 | - | 6 | 671 | |||
AK132783.1 | - | 6 | 705 | |||
AK145062.1 | - | 672 | ||||
AK146258.1 | - | 6 | 715 | |||
AK146374.1 | - | 6 | 716 | |||
AK156078.1 | - | 6 | 693 | |||
BC004718.1 | - | 6 | 689 | 3582426 |