Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pfkp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291071.1 | NP_001278000.1 | 22 | 2914 | ||
NM_019703.4 | NP_062677.1 | 22 | 2914 | |||
GenBank mRNA | AK047303.1 | - | 4 | 913 | ||
AK049841.1 | - | 22 | 2868 |