Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scoc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039137.3 | NP_001034226.1 | 4 | 497 | ||
NM_001285992.1 | NP_001272921.1 | 4 | 599 | |||
NM_019708.4 | NP_062682.1 | 4 | 602 | |||
GenBank mRNA | AF115778.1 | - | 4 | 420 | ||
AK002521.1 | - | 4 | 451 | |||
AK076085.1 | - | 4 | 395 | |||
AK167488.1 | - | 4 | 445 | |||
BC017629.1 | - | 4 | 421 | |||
BC085265.1 | - | 4 | 409 | 6391098 | ||
BC092071.1 | - | 4 | 405 | 4505111 | ||
BC108407.1 | - | 4 | 419 | |||
BC147069.1 | - | 4 | 490 | 9056075 | ||
CX241959.1 | - | 4 | 588 |