Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100169.1 | NP_001093639.1 | 3 | 565 | ||
NM_001100170.1 | NP_001093640.1 | 3 | 565 | |||
NM_006839.2 | NP_006830.2 | 3 | 565 | |||
XM_005264110.3 | XP_005264167.1 | 3 | 237 | |||
XM_005264113.1 | XP_005264170.1 | 3 | 233 | |||
XM_005264114.1 | XP_005264171.1 | 3 | 237 | |||
XM_011532497.2 | XP_011530799.1 | 3 | 237 | |||
XM_017003194.1 | XP_016858683.1 | 3 | 233 | |||
XM_017003195.1 | XP_016858684.1 | 3 | 233 | |||
GenBank mRNA | AK225780.1 | - | 3 | 232 | ||
AK296676.1 | - | 3 | 207 | |||
AK299014.1 | - | 3 | 228 | |||
AK300017.1 | - | 3 | 205 | |||
AK304197.1 | - | 3 | 242 | |||
AK309004.1 | - | 3 | 228 | |||
AK312251.1 | - | 3 | 215 | |||
AY232292.1 | - | 3 | 198 | |||
AY921637.1 | - | 3 | 213 | |||
BC002412.2 | - | 3 | 206 | 2964676 | ||
BC005983.1 | - | 3 | 257 | |||
BC107419.1 | - | 3 | 261 | |||
BX537369.1 | - | 3 | 268 | |||
CR456793.1 | - | 3 | 177 | |||
D21091.1 | - | 3 | 262 | |||
D21092.1 | - | 1 | 67 | |||
D21094.1 | - | 3 | 262 | |||
L42572.1 | - | 3 | 269 |