Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbms2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039080.1 | NP_001034169.1 | 13, 14 | 1489 | ||
NM_019711.2 | NP_062685.2 | 12, 13 | 1306 | |||
GenBank mRNA | AB026583.1 | - | 12, 13 | 1188 | ||
AK054482.1 | - | 12, 13 | 1219 | |||
AK147838.1 | - | 13, 14 | 1490 | |||
BC021627.1 | - | 12, 13 | 1200 | 4207338 |