Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019712.3 | NP_062686.1 | 5 | 903 | ||
GenBank mRNA | AK004114.1 | - | 5 | 867 | ||
AK205455.1 | - | 1 | 201 | |||
BC027396.1 | - | 5 | 852 | 4952242 | ||
BC056992.1 | - | 5 | 846 | 6818827 |