Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rassf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243748.1 | NP_001230677.1 | 6 | 1473 | ||
NM_019713.4 | NP_062687.1 | 5 | 1417 | |||
GenBank mRNA | AF132851.1 | - | 5 | 1250 | ||
AK146736.1 | - | 5 | 1216 | |||
AK177896.1 | - | 1 | 231 | |||
AK209367.1 | - | 1 | 60 | |||
BC002173.1 | - | 5 | 1344 | 3486727 | ||
BC052760.1 | - | 4 | 1787 |