Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019721.2 | NP_062695.2 | 4, 5 | 1046 | ||
GenBank mRNA | AF135789.1 | - | 2, 3 | 609 | ||
AK009492.1 | - | 1, 2 | 107 | |||
AK035061.1 | - | 3, 4 | 864 | |||
AK085189.1 | - | 4, 5 | 1033 | |||
AK089158.1 | - | 3, 4 | 817 | |||
AK151616.1 | - | 4, 5 | 1033 | |||
AK153113.1 | - | 4, 5 | 1033 | |||
BC012526.1 | - | 4, 5 | 1030 | 3981018 |