Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019727.2 | NP_062701.2 | 14 | 1587 | ||
GenBank mRNA | AB019214.1 | - | 14 | 1527 | ||
AF154120.1 | - | 14 | 1591 | |||
AK076198.1 | - | 6 | 571 | |||
AK149850.1 | - | 14 | 1499 | |||
AK154079.1 | - | 14 | 1500 | |||
AK155746.1 | - | 14 | 1475 | |||
AK161241.1 | - | 14 | 1568 | |||
AK167177.1 | - | 14 | 1542 | |||
AK167993.1 | - | 14 | 1482 | |||
AK168742.1 | - | 14 | 1580 | |||
BC066044.1 | - | 14 | 1544 | 6807363 | ||
BC066189.1 | - | 14 | 1478 | 30471747 |