Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rybp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019743.3 | NP_062717.2 | 5 | 1938 | ||
GenBank mRNA | BC053016.1 | - | 5 | 1938 | 6406584 | |
BC080287.1 | - | 5 | 2021 | 6410301 |