Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019746.4 | NP_062720.1 | 6 | 528 | ||
GenBank mRNA | AK007297.1 | - | 6 | 441 | ||
AK008632.1 | - | 6 | 542 | |||
BC048476.1 | - | 6 | 428 | 6773378 | ||
BC056167.1 | - | 6 | 420 | 5712632 | ||
BC092092.1 | - | 6 | 528 | 3492310 |