Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amotl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019764.2 | NP_062738.2 | 10 | 3457 | ||
GenBank mRNA | AF175967.1 | - | 10 | 2740 | ||
AF175968.1 | - | 8 | 3597 | |||
AK040912.1 | - | 10 | 3538 | |||
AK141348.1 | - | 10 | 3457 | |||
AK164279.1 | - | 10 | 3526 | |||
AK164819.1 | - | 11 | 3560 | |||
AK169419.1 | - | 10 | 3457 | |||
BC027824.1 | - | 10 | 3427 | 5323888 | ||
BC035201.1 | - | 10 | 3439 | |||
BC108411.1 | - | 10 | 3491 |