Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291229.1 | NP_001278158.1 | 8, 9 | 1580 | ||
NM_019765.5 | NP_062739.2 | 8, 9 | 1580 | |||
GenBank mRNA | AK220172.1 | - | 8, 9 | 1541 | ||
BC007191.1 | - | 8, 9 | 1520 | 3592614 | ||
BC029243.1 | - | 5, 6 | 702 | |||
BC052632.1 | - | 6, 7 | 986 |