Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291229.1 | NP_001278158.1 | 3 | 563 | ||
NM_019765.5 | NP_062739.2 | 3 | 563 | |||
GenBank mRNA | AK014540.1 | - | 1 | 276 | ||
AK029945.1 | - | 3 | 493 | |||
AK220172.1 | - | 3 | 524 | |||
BC007191.1 | - | 3 | 503 | 3592614 |