Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN2C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256494.1 | NP_001243423.1 | 3, 4 | 403 | ||
NM_001256495.1 | NP_001243424.1 | 3, 4 | 403 | |||
NM_001256496.1 | NP_001243425.1 | 3, 4 | 403 | |||
NM_006715.3 | NP_006706.2 | 3, 4 | 403 | |||
XM_005254384.2 | XP_005254441.1 | 3, 4 | 456 | |||
XM_017022186.1 | XP_016877675.1 | 2, 3 | 751 | |||
XM_017022187.1 | XP_016877676.1 | 2, 3 | 751 | |||
XM_017022188.1 | XP_016877677.1 | 3, 4 | 456 | |||
GenBank mRNA | AB209141.1 | - | 281 | |||
AF044414.2 | - | 3, 4 | 415 | |||
AK225145.1 | - | 3, 4 | 380 | |||
AK294892.1 | - | 3, 4 | 382 | |||
AK295265.1 | - | 3, 4 | 712 | |||
AK311065.1 | - | 2, 3 | 255 | |||
AL136876.1 | - | 3, 4 | 364 | |||
BC050550.1 | - | 3, 4 | 370 | 5529534 | ||
BC080191.1 | - | 3, 4 | 368 | 6149811 | ||
DC342919.1 | - | 3, 4 | 406 |