Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019773.2 | NP_062747.1 | 3 | 1293 | ||
GenBank mRNA | AB027290.1 | - | 3 | 1122 | ||
AK017301.1 | - | 3 | 1087 | |||
AK032133.1 | - | 3 | 1058 | |||
BC008160.1 | - | 3 | 1092 | 3964134 |